Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Test Tubes Racks

VWR®, Test Tubes Racks

Green, 40 (4×10), 25 mm, 123×301×94,5 mm
Catalog # 212-0420
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0420
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Green, 40 (4×10), 25 mm, 123×301×94,5 mm

Price
31,50 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    25 mm
  • No. of Holes:
    40 (4×10)
  • Colour:
    Green
  • Seq:
    39
  • W×D×H:
    123×301×94,5 mm