Order Entry
Italy
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Red, 40 (4×10), 25 mm, 123×301×94,5 mm
  212-0422
VWR®, Test Tubes Racks
  212-0422
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Red, 40 (4×10), 25 mm, 123×301×94,5 mm

 
31,50 €
   
1 * 1 pezzi
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    25 mm
  • No. of Holes:
    40 (4×10)
  • Colour:
    Red
  • Seq:
    41
  • W×D×H:
    123×301×94,5 mm