Funnels with short stem
25 mm, 200 mm
Catalog # SAMP8022P-200
Supplier: Sampling Systems
Jump to:
Product Details & Documents
These liquid funnels have PP construction. Crevice free.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Specifications
- Funnel O-Ø:200 mm
- Stem O-Ø:25 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



