Bottles, water sampling
500 ml, Undosed, bulk
Supplier: LP ITALIANA
Jump to:
Product Details & Documents
PP, translucent, square, with screw cap.
- Tamper evident screw caps
- Sterile
- Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Specifications
- Packed:bulk
- Capacity:500 ml
- Seq:03
- Dosage:Undosed