Order Entry
Italy
ContactUsLinkComponent
Areometer, DIN 12791, M50, 0,65-0,70:0,0010g/cm , ref. temp. 20°C, without thermometer max. 270mm long, suitable for government ver ification 1 * 1 items
Catalog # AMARH808002
Supplier:  Amarell
Areometer, DIN 12791, M50, 0,65-0,70:0,0010g/cm , ref. temp. 20°C, without thermometer max. 270mm long, suitable for government ver ification 1 * 1 items
Catalog # AMARH808002
Supplier:  Amarell
Frequently Bought Together