Order Entry
Italy
Orders LinkContactUsLinkComponent
Ribbons for label printer BBP™11

Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 4900 series, width 35 mm, BBP™11, 70 m, Black
Catalog # BRDY804792
Supplier:   Brady
Ribbons for label printer BBP™11
Catalog # BRDY804792
Supplier:  Brady
Ribbons for label printer BBP™11
Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 4900 series, width 35 mm, BBP™11, 70 m, Black

Price
12,40 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, 4900 series, width 35 mm
  • Length:
    70 m
  • Seq:
    01
  • For:
    BBP™11