Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, flj10986( np_060761, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00055277-Q01
Supplier:  Abnova
Recombinant protein, flj10986( np_060761, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00055277-Q01
Supplier:  Abnova