Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant protein, fbxo8( np_036312, 1 a.a. - 77 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00026269-Q01
:
Abnova
Recombinant protein, fbxo8( np_036312, 1 a.a. - 77 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00026269-Q01
:
Abnova
sds_ABNOH00026269-Q01.pdf
TDS_ABNOH00026269-Q01.pdf
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals