Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, yes1( aah48960, 1 a.a. - 80 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007525-Q01
Supplier:  Abnova
Recombinant protein, yes1( aah48960, 1 a.a. - 80 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007525-Q01
Supplier:  Abnova