Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, ca5b( np_009151, 2 a.a. - 90 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00011238-Q01
Supplier:  Abnova
Recombinant protein, ca5b( np_009151, 2 a.a. - 90 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00011238-Q01
Supplier:  Abnova