Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, lig3( np_039269, 823 a.a. - 922 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003980-Q01
Supplier:  Abnova
undefined
Recombinant protein, lig3( np_039269, 823 a.a. - 922 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003980-Q01
Supplier:  Abnova