Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, tceb2( np_009039, 9 a.a. - 118 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006923-Q01
Supplier:  Abnova
undefined
Recombinant protein, tceb2( np_009039, 9 a.a. - 118 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006923-Q01
Supplier:  Abnova