Order Entry
Italy
Orders LinkContactUsLinkComponent
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Phosphate test, 1,5-3-6-9-12-16-20-40-100 mg/l PO₄-P, Vanadato-molybdate, MCD, 300 Tests
Codice catalogo 1.18388.0001
Nome del fornitore:   Merck
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Codice catalogo 1.18388.0001
Nome del fornitore:  Merck
Numero CAS:  
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Phosphate test, 1,5-3-6-9-12-16-20-40-100 mg/l PO₄-P, Vanadato-molybdate, MCD, 300 Tests

Numero CAS:  
Price
347,00 €
Log in for availability
1 * 300 Tests
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. Ulteriori informazioni
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® with colour card comparator

By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.

Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.

Specifications

Specifications

  • Chemical method:
    Vanadato-molybdate
  • Pk:
    300 Tests
  • Description:
    Phosphate test
  • Type:
    MCD
  • Range:
    1,5-3-6-9-12-16-20-40-100 mg/l PO₄-P

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.