Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, cd8a( np_001759, 22 a.a. - 131 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Codice catalogo ABNOH00000925-Q01
Supplier:  Abnova
undefined
Recombinant protein, cd8a( np_001759, 22 a.a. - 131 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Codice catalogo ABNOH00000925-Q01
Supplier:  Abnova