Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, grik4( np_055434, 21 a.a. - 130 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Codice catalogo ABNOH00002900-Q01
Supplier:  Abnova
undefined
Recombinant protein, grik4( np_055434, 21 a.a. - 130 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Codice catalogo ABNOH00002900-Q01
Supplier:  Abnova