Recombinant protein, spink2( np_066937.1, 1 a.a. - 84 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00006691-P01
Supplier: Abnova
Recombinant protein, spink2( np_066937.1, 1 a.a. - 84 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00006691-P01
Supplier: Abnova
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection