Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, pbx2( np_002577.2, 1 a.a. - 430 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005089-P01
Supplier:  Abnova
Recombinant protein, pbx2( np_002577.2, 1 a.a. - 430 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005089-P01
Supplier:  Abnova