Order Entry
Italy
ContactUsLinkComponent
Recombinant protein, insl4( aah26254, 26 a.a. - 140 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003641-P01
Supplier:  Abnova
Recombinant protein, insl4( aah26254, 26 a.a. - 140 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003641-P01
Supplier:  Abnova