SP Bel-Art View Pack™ microscope slide holders
Refill slide holder pages, White
Catalog # 467-0137
Supplier: Bel-Art Products, a Part of SP
SP Bel-Art View Pack™ microscope slide holders
Catalog # 467-0137
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
Microscope slide holders, with ring binder.
- Standard 230×300 mm three-ring binder comes complete with ten 220×270 mm vinyl slide holder pages, enough for 160 slides
- Each page can hold 16 slides, in individual pockets with a white back and clear front
- Center flap keeps slides from falling out
- Fold the page at the center crease to retrieve slides
Refill View-Pack™ microscope slide holder pages – Pack of ten are available.
Specifications
- Colour:White
- Description:Refill slide holder pages
- Seq:02