SP Bel-Art View Pack™ microscope slide holders
Slide holder binder including ten pages, Blue/white, 160
467-0136
Microscope slide holders, with ring binder.
- Standard 230×300 mm three-ring binder comes complete with ten 220×270 mm vinyl slide holder pages, enough for 160 slides
- Each page can hold 16 slides, in individual pockets with a white back and clear front
- Center flap keeps slides from falling out
- Fold the page at the center crease to retrieve slides
Refill View-Pack™ microscope slide holder pages – Pack of ten are available.
- No. of slides:160
- Colour:Blue/white
- Description:Slide holder binder including ten pages
- Seq:01