Order Entry
Italy
Orders LinkContactUsLinkComponent
1,1'-Binaphthyl ≥98%

1,1'-Binaphthyl ≥98%

Glass bottle for solids, 1 g
Catalog # A16852.03
Irritant
1,1'-Binaphthyl ≥98%
Catalog # A16852.03
CAS Number:  604-53-5
1,1'-Binaphthyl ≥98%
1,1'-Binaphthyl ≥98%

Glass bottle for solids, 1 g

CAS Number:  604-53-5
Price
42,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 604-53-5 Molecular Formula: C20H14 Formula Weight: 254.33 Melting Point: 144-146° MDL No.: MFCD00041740

Synonyms:  1,1'-Binaphthalene
  • CAS Number:604-53-5
  • Formula:C₂₀H₁₄
  • MDL Number:MFCD00041740
  • Molecular Weight:254.33 g/mol
  • Boiling Pt:405 °C (1013 hPa)
  • Melting Pt:144…146 °C
  • Density:1.19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:210-070-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications