Order Entry
Italy
Orders LinkContactUsLinkComponent
Count plates, Petrifilm™
Count plates, Petrifilm™
3M™ Petrifilm™ Aqua Heterotrophic Count Plate (AQHC), 2×50 pieces, 100
Codice catalogo 710-0849
Nome del fornitore:   3M Food Safety
Count plates, Petrifilm™
Codice catalogo 710-0849
Nome del fornitore:  3M Food Safety
Numero CAS:  
Count plates, Petrifilm™
Count plates, Petrifilm™
3M™ Petrifilm™ Aqua Heterotrophic Count Plate (AQHC), 2×50 pieces, 100
Numero CAS:  
Price
107,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents
3M™ Petrifilm™ is an extremely cost-effective, practical and reliable method for testing equipment, raw materials, food products and their manufacturing environment; it also enables more frequent monitoring of critical control points. Fast and easy-to-approve results provide you with the information required to make the right decisions. The end result is an increase in product quality, improved process control and an overall reduction in operating costs.
  • Fast and accurate results in 48 hours
  • Space-saving
  • 3M™ Petrifilm™ plates can also be read in four seconds using the 3M™ Petrifilm™ plate reader
Specifications
Specifications
  • Pk:
    100
  • Description:
    3M™ Petrifilm™ Aqua Heterotrophic Count Plate (AQHC), 2×50 pieces