Order Entry
Italy
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate ≥99%

Methyl-4-hydroxybenzoate ≥99%

Plastic container, 5.000 g
Catalog # A14289.0I
Irritant
Methyl-4-hydroxybenzoate ≥99%
Catalog # A14289.0I
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate ≥99%
Methyl-4-hydroxybenzoate ≥99%

Plastic container, 5.000 g

CAS Number:  99-76-3
Price
555,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 99-76-3 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless pH: 5.8 Melting Point: 125-128° Boiling Point: ca 280° dec. Beilstein Reference: 509801 Merck Reference: 14,6107 MDL No.: MFCD00002352

Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,6107
  • Boiling Pt:∼280 °C (dec.)
  • Melting Pt:125…128 °C
  • Density:1.1208 g/cm³ (137 °C)
  • Storage Temperature:Ambient
  • EINECS:202-785-7
Specifications

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.