Order Entry
Italy
Orders LinkContactUsLinkComponent
Beakers, low form, with spout

Beakers, low form, with spout

50 ml, 55 mm, 40 mm
Catalog # WITG8.600.050
Supplier:   witeg Labortechnik
Beakers, low form, with spout
Catalog # WITG8.600.050
Supplier:  witeg Labortechnik
Beakers, low form, with spout
Beakers, low form, with spout

50 ml, 55 mm, 40 mm

Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

Quartz glass.

  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature

The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.

Specifications
  • Ext.Ø:
    40 mm
  • Capacity:
    50 ml
  • Height:
    55 mm