Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2,4-Difluorostyrene

Bottle, 5 g
Codice catalogo APOSPC6274-5G
Flammable
Irritant
Nome del fornitore:   Apollo Scientific
2,4-Difluorostyrene
Codice catalogo APOSPC6274-5G
Nome del fornitore:  Apollo Scientific
Numero CAS:  399-53-1
2,4-Difluorostyrene

Bottle, 5 g

Numero CAS:  399-53-1
Price
157,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,4-Difluoro-1-ethenylbenzene, 2,4-Difluoro-1-vinylbenzene
  • CAS Number:399-53-1
  • Formula:C₈H₆F₂
  • MDL Number:MFCD00142824
  • Molecular Weight:140.13 g/mol
  • Boiling Pt:60…62 °C (10 mm Hg)
  • Storage Temperature:Refrigerator
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications