Interferin-hts sirna transfection reagent is especially developed for high troughput screening (hts) applications providing great si lencing efficiency, excellent reproducibility and high cell viability with very low amounts of reagent. suitable for 1 * 1,5 ml
Catalog # 410-015
Supplier: Polyplus-transfection
Frequently Bought Together