Order Entry
Italy
Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96 Well plate, Translucent, 96, +
Catalog # RATI6018123
Supplier:   RATIOLAB
Microplates, 96-well
Catalog # RATI6018123
Supplier:  RATIOLAB
Microplates, 96-well
Microplates, 96-well
96 Well plate, Translucent, 96, +
Price
110,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents
Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Colour:
    Translucent
  • Description:
    96 Well plate
  • Cat. No.:
    RATI6018123
  • Model:
    Micro Test Plates, volume ∼300 μl
  • Well profile:
    U-bottom