Order Entry
Italy
Orders LinkContactUsLinkComponent
Serological pipettes

Serological pipettes

25 ml, 0,2 ml, +, Red, Individually
Catalog # RATI3125011
Supplier:   RATIOLAB
Serological pipettes
Catalog # RATI3125011
Supplier:  RATIOLAB
Serological pipettes
Serological pipettes

25 ml, 0,2 ml, +, Red, Individually

Price
156,00 €
Log in for availability
1 * 200 pezzi
Product Details & Documents

Disposable pipettes made from clear polystyrene (PS).

  • Free from RNase, DNase, DNA and pyrogens
  • Sterile, individually packed
  • From one mould – hence more accurate and stable and with minimal residual wetting
  • Permanent black embossed graduations
  • Colour coded cotton wool plug for convenient identification
  • Negative graduation for additional capacity
  • 5, 10 and 25 ml pipettes with scales for dispensed and the residual volumes
Specifications
  • Colour code:
    Red
  • Packed:
    Individually
  • Sterile:
    +
  • Capacity:
    25 ml
  • Division:
    0,2 ml