Base materials, Millipore®
Proteose peptone from animal tissue, for antibiotic production, growth of fastidious microorganisms and production of bacterial toxins, 250 g
Catalog # SIALP0431-250G
Supplier: Merck
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:250 g
- Description:Proteose peptone from animal tissue, for antibiotic production, growth of fastidious microorganisms and production of bacterial toxins