Order Entry
Italy
Orders LinkContactUsLinkComponent
Filling cannulae
Filling cannulae
Filling cannulae, G 18, 50 mm
Catalog # MURRM718/03
Supplier:   MURRAY S
Filling cannulae
Catalog # MURRM718/03
Supplier:  MURRAY S
Filling cannulae
Filling cannulae, G 18, 50 mm
Price
31,80 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
Specifications
  • Gauge:
    G 18
  • Description:
    Filling cannulae
  • Length:
    50 mm
  • Seq:
    004