Filling cannulae
Filling cannulae, G 14, 150 mm
Catalog # MURRM714/08
Supplier: MURRAY S
Jump to:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Datasheet
Specifications
- Gauge:G 14
- Description:Filling cannulae
- Length:150 mm
- Seq:017