Order Entry
Italy
Orders LinkContactUsLinkComponent
Filling cannulae
Filling cannulae
Filling cannulae, G 14, 76 mm
Catalog # MURRM714/05
Supplier:   MURRAY S
Filling cannulae
Catalog # MURRM714/05
Supplier:  MURRAY S
Filling cannulae
Filling cannulae, G 14, 76 mm
Price
44,30 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
Specifications
  • Gauge:
    G 14
  • Description:
    Filling cannulae
  • Length:
    76 mm
  • Seq:
    007