Hypodermic needles
Hypodermic, sharp, 16, 50 mm
Catalog # MURRM616/03
Supplier: MURRAY S
Jump to:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifications
- Gauge:16
- Description:Hypodermic, sharp
- Length:50 mm