Hypodermic needles
Hypodermic, sharp, 14, 76 mm
Catalog # MURRM614/05
Supplier: MURRAY S
Jump to:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifications
- Gauge:14
- Description:Hypodermic, sharp
- Length:76 mm