Order Entry
Italy
Orders LinkContactUsLinkComponent
Biotin kits, HOOK™
Biotin kits, HOOK™
Biotin-Hydrazide kit, 1 KIT
Catalog # BS-18
Supplier:   G-Biosciences
Biotin kits, HOOK™
Catalog # BS-18
Supplier:  G-Biosciences
CAS Number:  
Biotin kits, HOOK™
Biotin kits, HOOK™
Biotin-Hydrazide kit, 1 KIT
CAS Number:  
Price
335,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
HOOK™ Biotin Reagent that conjugates to proteins through amines, sulfhydryls, carboxyls or carbohydrates. The amine and sulfhydryl coupling HOOK™ Biotin Reagents couple directly to the protein through their reactive groups, however the carboxyl coupling HOOK™ Biotin Reagents require a carbodiimide crosslinker and the carbohydrate coupling HOOK™ Biotin Reagents require carbohydrate oxidation before coupling.
  • Optimizer Buffer™ for improved coupling efficiency
  • SpinOUT™ gel filtration columns for rapid (<10 minutes) purification
  • Biotin assay reagents to determine level of biotin incorporation
  • Labels 1 to 10 mg protein/reaction
  • Suitable for 10 coupling reactions
The kits include EDC as the carbodiimide crosslinker in the carboxyl coupling kits and sodium meta-periodate for carbohydrate oxidation in the carbohydrate coupling kits. Kit also contains a specific Optimizer Buffer™ that provides the optimal reaction conditions for each HOOK™ Biotin Reagent.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Biotin-Hydrazide kit
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-V, 5X
    Optimizer Buffer™-V, 5X
    BKC-08
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-III, 5X
    Optimizer Buffer™-III, 5X
    BKC-06
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-IV, 5X
    Optimizer Buffer™-IV, 5X
    BKC-07
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-VI, 5X
    Optimizer Buffer™-VI, 5X
    BKC-09
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-II,5X
    Optimizer Buffer™-II,5X
    BKC-05
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Optimizer Buffer™-I, 5X
    Optimizer Buffer™-I, 5X
    BKC-04
    2
    Log in for availability
    1 * 2 pezzi - 149,00 €