Order Entry
Italy
Orders LinkContactUsLinkComponent
dNTP mix
Deoxynucleotide Mix, 10 mM solution, 0,5 ml, 5 µmol
  786-443
dNTP mix
  786-443
 :  
dNTP mix
dNTP mix
Deoxynucleotide Mix, 10 mM solution, 0,5 ml, 5 µmol
 :  
59,50 €
1 * 5 umol
Aqueous mixture of dATP, dCTP, dGTP and dTTP each at a final concentration of 10 or 25 mM.
  • Pk:
    5 µmol
  • Description:
    Deoxynucleotide Mix, 10 mM solution, 0,5 ml