Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,4-Dimethylhexane ≥97%

Glass bottle for solids, 25 g
Catalog # H53498.14
Flammable
Health
Irritant
Environment
3,4-Dimethylhexane ≥97%
Catalog # H53498.14
CAS Number:  583-48-2
3,4-Dimethylhexane ≥97%
3,4-Dimethylhexane ≥97%

Glass bottle for solids, 25 g

CAS Number:  583-48-2
Price
346,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:583-48-2
  • Formula:C₈H₁₈
  • MDL Number:MFCD00039927
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:119 °C
  • Melting Pt:–64 °C
  • Density:0.72
  • Flash Pt:6 °C (43 °F)
  • Storage Temperature:Ambient
  • EINECS:209-504-7
  • UN:1262
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications