Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™
AmphiSpheres™, resins, 40 RAM, 0,4 mmol/g, 1 kg, 75 - 150 µm, 1
Catalog # VARIPL3867-6764
Supplier: VARIAN
Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™
Catalog # VARIPL3867-6764
Supplier: VARIAN
CAS Number:
Jump to:
Product Details & Documents
Strongly basic anion exchanger, Cl⁻ form. Moist, light yellow spherules.
Specifications
Specifications
- Particle Size:75 - 150 µm
- Pk:1
- Description:AmphiSpheres™, resins, 40 RAM, 0,4 mmol/g, 1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



