Order Entry
Italy
Orders LinkContactUsLinkComponent
Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™

Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™

AmphiSpheres™, resins, 20 HMP, 0,7 mmol/g, 1 kg, 75 - 150 µm, 1
Catalog # VARIPL3863-6762
Supplier:   VARIAN
Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™
Catalog # VARIPL3863-6762
Supplier:  VARIAN
CAS Number:  
Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™

AmphiSpheres™, resins, 20 HMP, 0,7 mmol/g, 1 kg, 75 - 150 µm, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

Strongly basic anion exchanger, Cl⁻ form. Moist, light yellow spherules.

Specifications

Specifications

  • Particle Size:
    75 - 150 µm
  • Pk:
    1
  • Description:
    AmphiSpheres™, resins, 20 HMP, 0,7 mmol/g, 1 kg

Specifications