Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™
AmphiSpheres™, resins, 20 HMP, 0,7 mmol/g, 100 g, 75 - 150 µm, 1
Catalog # VARIPL3863-4762
Supplier: VARIAN
Amphiathic resin for solid phase peptide synthesis, AmphiSpheres™
Catalog # VARIPL3863-4762
Supplier: VARIAN
CAS Number:
Jump to:
Product Details & Documents
Strongly basic anion exchanger, Cl⁻ form. Moist, light yellow spherules.
Specifications
Specifications
- Dimensioni particelle:75 - 150 µm
- Conf:1
- Descrizione:AmphiSpheres™, resins, 20 HMP, 0,7 mmol/g, 100 g



