Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 30, 8 µm, 7,50 mm, 300 mm
Catalog # VARIPL1120-6830
Supplier:   VARIAN
Columns and standards for GPC/SEC
Catalog # VARIPL1120-6830
Supplier:  VARIAN
Columns and standards for GPC/SEC
Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 30, 8 µm, 7,50 mm, 300 mm

Price
2 850,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Ø int.:
    7,50 mm
  • Particle Size:
    8 µm
  • Description:
    Aqueous SEC of polymers, PL-aquagel-OH 30
  • Length:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.