Order Entry
Italy
ContactUsLinkComponent
B#CHI Kjeldahl-System K-370 (230 V, 50/60 Hz) The AutoKjeldahl Unit K-370 is a compact, fully automatic distillation, titration and evaluation system in an acid- and alkali-resista 1 * 1 items
Catalog # BUCH043110
Supplier:  BUCHI
B#CHI Kjeldahl-System K-370 (230 V, 50/60 Hz) The AutoKjeldahl Unit K-370 is a compact, fully automatic distillation, titration and evaluation system in an acid- and alkali-resista 1 * 1 items
Catalog # BUCH043110
Supplier:  BUCHI
Frequently Bought Together