Order Entry
Italy
Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
White, poly-D-lysine, lid with condensation rings, 25 - 340 µl, -, 392 µl
Catalog # GREI655944BE
Microplates, 96-well
Catalog # GREI655944BE
Supplier:  
Microplates, 96-well
Microplates, 96-well
White, poly-D-lysine, lid with condensation rings, 25 - 340 µl, -, 392 µl
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PLATE 96W WHITE/µCL FLAT PDL LID N-ST 1 * 20 items Catalog #392-0027
Product 392-0027 is a replacement for GREI655944BE but varies by Brand, Packaging or Quantity.
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications
  • Recommended working volume:
    25 - 340 µl
  • Sterile:
    -
  • Description:
    White, poly-D-lysine, lid with condensation rings
  • Seq:
    16
  • Total volume:
    392 µl
  • Cat. No.:
    GREI655944BE