Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Small reaction vials, Reacti-Vial™

Vial, inside Ø 11 mm, thread style 425-13, 1,00 ml, 13×45 mm, Amber, Yes
  HYPETS-13097
Small reaction vials, Reacti-Vial™
  HYPETS-13097
Small reaction vials, Reacti-Vial™
Small reaction vials, Reacti-Vial™

Vial, inside Ø 11 mm, thread style 425-13, 1,00 ml, 13×45 mm, Amber, Yes

 
232,00 €
   
1 * 12 pezzi
 

These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.

  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds

These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solvent separations.

 : Vials are supplied complete with open-top screw caps and teflon/rubber laminated discs.

 
  • Cap included:
    Yes
  • Ø×H:
    13×45 mm
  • Capacity:
    1,00 ml
  • Colour:
    Amber
  • Description:
    Vial, inside Ø 11 mm, thread style 425-13
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.