Order Entry
Italy
ContactUsLinkComponent
Column, TSK-gel, gel permeation column, SuperH4000, 3 µm, inner diameter 6mm, length 15cm, stainless steel. 1 * 1 items
Catalog # TOYO0017994
Supplier:  TOSOH Bioscience
Column, TSK-gel, gel permeation column, SuperH4000, 3 µm, inner diameter 6mm, length 15cm, stainless steel. 1 * 1 items
Catalog # TOYO0017994
Supplier:  TOSOH Bioscience
Frequently Bought Together