Order Entry
Italy
Orders LinkContactUsLinkComponent
Moria Fine scissors

Moria Fine scissors

Sharp/sharp, straight tips, 105 mm, 25 mm
Catalog # FSCI14370-22
Marketsource
Supplier:   FINE SCIENCE TOOLS
Moria Fine scissors
Catalog # FSCI14370-22
Supplier:  FINE SCIENCE TOOLS
Marketsource
Moria Fine scissors
Moria Fine scissors

Sharp/sharp, straight tips, 105 mm, 25 mm

Price
204,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These scissors are made of inox.

Specifications
  • Type:
    Sharp/sharp, straight tips
  • Length:
    105 mm
  • Seq:
    01
  • Blade length:
    25 mm