Order Entry
Italy
Orders LinkContactUsLinkComponent
Moria Fine scissors

Moria Fine scissors

Sharp/sharp, curved tips, 105 mm, 25 mm
Catalog # FSCI14370-23
Marketsource
Supplier:   FINE SCIENCE TOOLS
Moria Fine scissors
Catalog # FSCI14370-23
Supplier:  FINE SCIENCE TOOLS
Marketsource
Moria Fine scissors
Moria Fine scissors

Sharp/sharp, curved tips, 105 mm, 25 mm

Price
209,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These scissors are made of inox.

Specifications
  • Type:
    Sharp/sharp, curved tips
  • Length:
    105 mm
  • Seq:
    02
  • Blade length:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.