Shodex GPC KF-G 8B - polymer based organic HPLC guard column - SEC mode - THF - preparative - ID: 8mm length: 50mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 15 - Pore Size (A): - - Exclusion Limit 1 * 1 items
SHOWF6700406
: Resonac
Frequently Bought Together