Order Entry
Italy
Orders LinkContactUsLinkComponent
DNA prep for PCR - tissue, Extracta™

DNA prep for PCR - tissue, Extracta™

Extracta™ DNA prep for PCR - tissue, 250 mL
Catalog # 733-2212
Supplier:   Quantabio
DNA prep for PCR - tissue, Extracta™
Catalog # 733-2212
Supplier:  Quantabio
CAS Number:  
DNA prep for PCR - tissue, Extracta™

Extracta™ DNA prep for PCR - tissue, 250 mL

CAS Number:  
Price
1 330,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

The Extracta™ DNA prep for PCR - tissue consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva.

  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, high resolution melt analysis, endpoint PCR and other applications
  • Works with wide range of tissue
  • Optional 10 minute protocol for high throughput applications

The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Extracta™ DNA prep for PCR - tissue
  • Cat. No.:
    733-2212
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Extraction for direct PCR
  • Description:
    Extracta™ DNA prep for PCR - tissue
  • Sample type:
    Cultured cells|Human/animal cells and tissues

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.