Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Columns, INdEX™

100/500, Glass, 100×500 mm
  18-1104-15
 :   Cytiva
Columns, INdEX™
  18-1104-15
 :  Cytiva
Columns, INdEX™
Columns, INdEX™

100/500, Glass, 100×500 mm

 
 
   
1 * 1 pezzi
 

INdEX™ columns are easy-to-use, general purpose, glass columns well-suited for applications such as process development and diagnostics production. These columns are characterised by their simple design and the novel, axial compression packing method that yields a densely packed bed in under 10 minutes.

  • Scalable from inner diameters of 70 to 200 mm and bed volumes from 0,1 to 25 L
  • For use with an array of techniques and chromatography media, especially Sepharose Fast Flow
  • Proven distribution system
  • Dynamic axial compression yields densely packed, highly efficient beds
  • Materials include electropolished stainless steel, borosilicate glass and polymers
  • Packing devices for longer bed heights
 
  • Description:
    100/500
  • Column hardware:
    Glass
  • Bed dimensions:
    100×500 mm
  • Cat. No.:
    18-1104-15
  • Max. pressure:
    3 bar
Accessories

 

  •  
    Description
     
     
  • 18-1012-69
    Connector, 25 mm TC, I.Ø 22 mm, 51 mm TC, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
       
    1 * 1 pezzi - 375,00 €

 

  •  
    Description
     
     
  • 18-1031-09
    Connector, 25 mm TC-M6 threaded, for INdEX™ 70 and INdEX™ 100 columns
       
    1 * 2 pezzi - 100,00 €

 

  •  
    Description
     
     
  • 18-1104-29
    T-junction, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
       
    1 * 1 pezzi - 572,00 €

 

  •  
    Description
     
     
  • 18-5758-01
    Valves, 4-port, 4-way, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
       
    1 * 1 pezzi - 1 850,00 €

 

  •  
    Description
     
     
  • 18-0005-45
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
       
    1 * 1 pezzi - 437,00 €